Learn More
Cayman Chemical Indole-3-acetonItrIle 5g
SDP
Supplier:  Cayman Chemical 387515 G
Encompass_Preferred
An indole alkaloid with antibacterial activity; reduces biofilm formation by E. coli and P. aeruginosa at 100 UG/ml; levels are increased in P. brassicae-infected cabbage roots
Catalog No. 50-291-7654