Learn More
Medchemexpress LLC Indole-3-acetyl glutamate | 57105-48-3 | MFCD16036311 | 99.4% | 304.30 g·mol⁻¹ | C15H16N2O5 | 10 MG
SDP
Catalog No. 5000285262
Encompass_Preferred
Indole-3-acetyl glutamate is a conjugated form of the plant hormone indole-3-acetic acid (IAA) used in plant physiology and hormone research to study IAA transport, storage, and homeostasis. Supplied as a purified solid for research applications.
  • High purity suitable for analytical and biological research (99.35%).
  • Available in small, research-friendly quantities, including 10 mg.
  • Solid, off-white to light brown appearance for straightforward handling.
  • Stable for long-term storage as a powder at -20°C.
  • Provided with chemical identifiers (CAS, formula, SMILES) for accurate identification.
Catalog No. 50-002-85262 Supplier Medchemexpress LLC Supplier No. HYW58826310MG
Indole-3-acetyl glutamate is a conjugated form of the plant hormone indole-3-acetic acid (IAA) used in plant physiology and hormone research to study IAA transport, storage, and homeostasis. Supplied as a purified solid for research applications.
  • High purity suitable for analytical and biological research (99.35%).
  • Available in small, research-friendly quantities, including 10 mg.
  • Solid, off-white to light brown appearance for straightforward handling.
  • Stable for long-term storage as a powder at -20°C.
  • Provided with chemical identifiers (CAS, formula, SMILES) for accurate identification.