Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Indole-3-acetylglycine | 13113-08-1 | MFCD00079392 | 95.0% | 232.24 | C12H12N2O3 | 250 MG
SDP

Catalog No. 5000280658
Encompass_Preferred

Indole-3-acetylglycine is an indole-derived biochemical reagent used in laboratory research and assay workflows. It is supplied as a solid for use in biochemical assays and sample preparation. Users should consult the certificate of analysis for lot-specific purity and handling information.

  • Used as a biochemical assay reagent in research applications.
  • Molecular formula C12H12N2O3.
  • Molecular weight 232.24 g/mol.
  • CAS number 13113-08-1.
  • Available in multiple package sizes; verify pack size with the supplier.
  • Refer to certificate of analysis for lot-specific purity and storage instructions.

Catalog No. 50-002-80658 Supplier Medchemexpress LLC Supplier No. HYW317414250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Indole-3-acetylglycine is an indole-derived biochemical reagent used in laboratory research and assay workflows. It is supplied as a solid for use in biochemical assays and sample preparation. Users should consult the certificate of analysis for lot-specific purity and handling information.

  • Used as a biochemical assay reagent in research applications.
  • Molecular formula C12H12N2O3.
  • Molecular weight 232.24 g/mol.
  • CAS number 13113-08-1.
  • Available in multiple package sizes; verify pack size with the supplier.
  • Refer to certificate of analysis for lot-specific purity and storage instructions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.