Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Indole-3-amidoxime | 95649-37-9 | MFCD00460181 | 96.3% | 175.19 g/mol | C9H9N3O | 1 G
SDP

Catalog No. 5000284019
Encompass_Preferred

Indole-3-amidoxime is a small-molecule synthetic intermediate used in pharmaceutical and medicinal chemistry research. It is supplied as an off-white to light-yellow solid for laboratory use only and is not intended for human or clinical applications.

  • Used as a synthetic intermediate in medicinal chemistry and drug development
  • Molecular formula C9H9N3O and molecular weight 175.19 g/mol
  • CAS number 95649-37-9 for unambiguous identification
  • High purity (96.3%) suitable for research applications
  • Off-white to light-yellow solid, easy to handle and weigh
  • Recommended storage: powder at -20°C (stable up to 3 years) or 4°C (up to 2 years)
  • Available in small lab-scale pack sizes for synthesis and screening

Catalog No. 50-002-84019 Supplier Medchemexpress LLC Supplier No. HYW5909111G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Indole-3-amidoxime is a small-molecule synthetic intermediate used in pharmaceutical and medicinal chemistry research. It is supplied as an off-white to light-yellow solid for laboratory use only and is not intended for human or clinical applications.

  • Used as a synthetic intermediate in medicinal chemistry and drug development
  • Molecular formula C9H9N3O and molecular weight 175.19 g/mol
  • CAS number 95649-37-9 for unambiguous identification
  • High purity (96.3%) suitable for research applications
  • Off-white to light-yellow solid, easy to handle and weigh
  • Recommended storage: powder at -20°C (stable up to 3 years) or 4°C (up to 2 years)
  • Available in small lab-scale pack sizes for synthesis and screening

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.