Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Indole-3-amidoxime | 95649-37-9 | MFCD00460181 | 96.3% | 175.19 g/mol | C9H9N3O | 250 MG
SDP

Catalog No. 5000284281
Encompass_Preferred

Indole-3-amidoxime is a small-molecule synthetic intermediate used in pharmaceutical and medicinal chemistry for building and modifying indole-based heterocycles. It is supplied as a powder with documented solubility and storage conditions for research use.

  • Synthetic intermediate suitable for pharmaceutical synthesis.
  • Chemical formula C9H9N3O.
  • Molecular weight 175.19 g/mol.
  • Typical purity 96.28%.
  • Appearance: powder for laboratory use.
  • Solubility: soluble in DMSO (reported 35 mg/mL; may require ultrasonic).
  • Storage: powder stored at -20°C (long term) or 4°C (short term).

Catalog No. 50-002-84281 Supplier Medchemexpress LLC Supplier No. HYW590911250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Indole-3-amidoxime is a small-molecule synthetic intermediate used in pharmaceutical and medicinal chemistry for building and modifying indole-based heterocycles. It is supplied as a powder with documented solubility and storage conditions for research use.

  • Synthetic intermediate suitable for pharmaceutical synthesis.
  • Chemical formula C9H9N3O.
  • Molecular weight 175.19 g/mol.
  • Typical purity 96.28%.
  • Appearance: powder for laboratory use.
  • Solubility: soluble in DMSO (reported 35 mg/mL; may require ultrasonic).
  • Storage: powder stored at -20°C (long term) or 4°C (short term).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.