Learn More
Medchemexpress LLC Indole-3-amidoxime | 95649-37-9 | MFCD00460181 | 96.3% | 175.19 g/mol | C9H9N3O | 500 MG
SDP
Catalog No. 5000285266
Encompass_Preferred
Indole-3-amidoxime is an indole-derived synthetic intermediate commonly used in pharmaceutical research and medicinal chemistry for building and modifying heterocyclic compounds.
  • Indole-derived amidoxime intermediate suitable for synthetic transformations.
  • Molecular formula C9H9N3O and molecular weight 175.19 g/mol.
  • Typical purity approximately 96.3% as supplied for research use.
  • Available in multiple pack sizes, including a 500 mg quantity for small-scale synthesis.
  • Useful for drug intermediate development and method optimization in medicinal chemistry.
Catalog No. 50-002-85266 Supplier Medchemexpress LLC Supplier No. HYW590911500MG
Indole-3-amidoxime is an indole-derived synthetic intermediate commonly used in pharmaceutical research and medicinal chemistry for building and modifying heterocyclic compounds.
  • Indole-derived amidoxime intermediate suitable for synthetic transformations.
  • Molecular formula C9H9N3O and molecular weight 175.19 g/mol.
  • Typical purity approximately 96.3% as supplied for research use.
  • Available in multiple pack sizes, including a 500 mg quantity for small-scale synthesis.
  • Useful for drug intermediate development and method optimization in medicinal chemistry.