Learn More
Cayman Chemical Indole-3-butyric Acid, Purity: ≥98%, CAS: 133-32-4, 100 mg
SDP
Catalog No. 502986871
Encompass_Preferred
A plant growth regulator; enhances root formation and improves S. rebaudiana plant survival; increases the amount of differentiated tissue and solasodine production by S. elaeagnifolium seedlings in a dose-dependent manner
Catalog No. 50-298-6871 Supplier Cayman Chemical Supplier No. 22591100 MG
A plant growth regulator; enhances root formation and improves S. rebaudiana plant survival; increases the amount of differentiated tissue and solasodine production by S. elaeagnifolium seedlings in a dose-dependent manner