Learn More
Medchemexpress LLC Indole-3-butyric acid | 133-32-4 | MFCD00005664 | 99.95% | 203.24 | 1 ML
SDP
Catalog No. 5000427319
Encompass_Preferred
Indole-3-butyric acid is an endogenous metabolite that functions as a plant growth auxin and a good rooting agent. It promotes the rooting of herbs and woody ornamental plants and can improve fruit rates. This compound is an auxin precursor that is converted to indole-3-acetic acid (IAA) through a peroxisomal β-oxidation process.
  • Functions as a plant growth auxin.
  • Acts as an effective rooting agent.
  • Promotes rooting in herbs and woody ornamental plants.
  • Can improve the fruit rate.
  • Is an auxin precursor.
  • Converts to indole-3-acetic acid (IAA) via peroxisomal β-oxidation.
Catalog No. 50-004-27319 Supplier Medchemexpress LLC Supplier No. HYN018610MM/1ML
Indole-3-butyric acid is an endogenous metabolite that functions as a plant growth auxin and a good rooting agent. It promotes the rooting of herbs and woody ornamental plants and can improve fruit rates. This compound is an auxin precursor that is converted to indole-3-acetic acid (IAA) through a peroxisomal β-oxidation process.
  • Functions as a plant growth auxin.
  • Acts as an effective rooting agent.
  • Promotes rooting in herbs and woody ornamental plants.
  • Can improve the fruit rate.
  • Is an auxin precursor.
  • Converts to indole-3-acetic acid (IAA) via peroxisomal β-oxidation.