Learn More
Medchemexpress LLC HY-N0186 5g Medchemexpress, Indole-3-butyric acid CAS:133-32-4 Purity:>98%
SDP
Catalog No. 501933453
Encompass_Preferred
Medchemexpress, HY-N0186 5g Indole-3-butyric acid CAS:133-32-4 Indole-3-butyric acid (3-indolebutyric acid; IBA) is a plant growth auxin and a good rooting agent. It can promote herbs and woody ornamental plant rooting and used for improving fruit rate. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3453 Supplier Medchemexpress LLC Supplier No. HYN01865G
Medchemexpress, HY-N0186 5g Indole-3-butyric acid CAS:133-32-4 Indole-3-butyric acid (3-indolebutyric acid; IBA) is a plant growth auxin and a good rooting agent. It can promote herbs and woody ornamental plant rooting and used for improving fruit rate. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.