Learn More
Indole-3-butyric acid, 98%, Thermo Scientific Chemicals
Description
Indole-3-butyric acid is auxin-family plant hormone (phytohormone). IBA is thought to be a precursor of indole-3-acetic acid (IAA) the most abundant and the basic auxin natively occurring and functioning in plants. IAA generates the majority of auxin effects in intact plants, and is the most potent native auxin.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indole-3-butyric acid is auxin-family plant hormone (phytohormone). IBA is thought to be a precursor of indole-3-acetic acid (IAA) the most abundant and the basic auxin natively occurring and functioning in plants. IAA generates the majority of auxin effects in intact plants, and is the most potent native auxin.
Solubility
Soluble in water(0.25g/L).
Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Melting Point | 121°C to 125°C |
Assay Percent Range | 98% |
Quantity | 5 g |
UN Number | UN2811 |
Beilstein | 171120 |
Merck Index | 14,4965 |
Solubility Information | Soluble in water(0.25g/L). |
Formula Weight | 203.24 |
Percent Purity | 98% |
Chemical Name or Material | Indole-3-butyric Acid |
Safety and Handling
Hazard Category | H301-H315-H319-H335 |
Hazard Statement | GHS H Statement H301-H315-H319-H335 |
Precautionary Statement | P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S. |
EINECSNumber | 205-101-5 |
RTECSNumber | NL5250000 |
TSCA | Yes |
storageNote1 | Air sensitive |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.