Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Indole-3-Butyric Acid , MP Biomedicals, LLC

Catalog No. MP210204250 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
50 g
Packaging:
Glass
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MP210204250 50 g Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP210204250 Supplier MP Biomedicals, Inc Supplier No. 0210204250
Add to Cart
Add to Cart

Indole-3-butyric has a purity of 99.2% acid is used as plant growth hormone.

  • A naturally occurring phytohormone auxin (plant growth regulator)
  • Promotes root formation in cuttings, but does not affect ethylene levels.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 133-32-4
Molecular Formula C12H13NO2
Molecular Weight (g/mol) 203.24
InChI Key JTEDVYBZBROSJT-UHFFFAOYSA-N
Synonym indole-3-butyric acid, 3-indolebutyric acid, 4-1h-indol-3-yl butanoic acid, indolebutyric acid, hormodin, 1h-indole-3-butanoic acid, seradix, indole-3-butanoic acid, jiffy grow, 4-indol-3-yl butyric acid
PubChem CID 8617
ChEBI CHEBI:33070

Specifications

Melting Point 118°C to 126°C
Color White
Packaging Glass
Quantity 50 g
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.