Learn More
Cayman Chemical Indole-3-carbInol 1g
SDP
Catalog No. 502953107
Encompass_Preferred
A natural phytochemical that has anti-cancer effects, at least in part through inhibition of NF-κB and Akt signaling pathways
Catalog No. 50-295-3107 Supplier Cayman Chemical Supplier No. 113251 G
A natural phytochemical that has anti-cancer effects, at least in part through inhibition of NF-κB and Akt signaling pathways