Learn More
Medchemexpress LLC HY-W007376 100mg Medchemexpress, Indole-3-carboxaldehyde CAS:487-89-8 Purity:>98%
SDP
Catalog No. 501970649
Encompass_Preferred
Medchemexpress, HY-W007376 100mg Indole-3-carboxaldehyde CAS:487-89-8 Indole-3-carboxaldehyde (3-Formylindole), a cabbage extract, is the product of the oxidative degradation of indole-3-acetic acid (IAA) by crude enzyme preparations from etiolated pea seedlings. Indole-3-carboxaldehyde (3-Formylindole) is a biochemical used to prepare analogs of the indole phytoalexin cyclobrassinin. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0649 Supplier Medchemexpress LLC Supplier No. HYW007376100MG
Medchemexpress, HY-W007376 100mg Indole-3-carboxaldehyde CAS:487-89-8 Indole-3-carboxaldehyde (3-Formylindole), a cabbage extract, is the product of the oxidative degradation of indole-3-acetic acid (IAA) by crude enzyme preparations from etiolated pea seedlings. Indole-3-carboxaldehyde (3-Formylindole) is a biochemical used to prepare analogs of the indole phytoalexin cyclobrassinin. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.