Learn More
Cayman Chemical Indole-3-carboxaldehyde 10g
SDP
Catalog No. 502991736
Encompass_Preferred
An active tryptophan metabolite and synthetic intermediate; increases colonic production of IL-22 and restores colonization resistance to C. albicans infection in a wild-type, but not Ahr-/-, mouse model of mucosal candidiasis at 18 mg/kg; reduces intestinal mucosal damage in a mouse model of DSS-induced colitis
Catalog No. 50-299-1736 Supplier Cayman Chemical Supplier No. 3132810 G
An active tryptophan metabolite and synthetic intermediate; increases colonic production of IL-22 and restores colonization resistance to C. albicans infection in a wild-type, but not Ahr-/-, mouse model of mucosal candidiasis at 18 mg/kg; reduces intestinal mucosal damage in a mouse model of DSS-induced colitis