Learn More
Medchemexpress LLC Indole-3-carboxaldehyde-13C3 | 1093452-52-8 | 146.15 | 5 MG
SDP
Catalog No. 5000460264
Encompass_Preferred
Indole-3-carboxaldehyde-13C3 (3-Formylindole-13C3) is a 13C labeled Indole-3-carboxaldehyde. It is a biochemical used to prepare analogs of the indole phytoalexin cyclobrassinin. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, primarily as tracers for quantitation during the drug development process. Deuteration has gained attention due to its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • For research use only
  • Incorporates stable heavy isotopes for quantitation in drug development
Catalog No. 50-004-60264 Supplier Medchemexpress LLC Supplier No. HYW007376S5MG
Indole-3-carboxaldehyde-13C3 (3-Formylindole-13C3) is a 13C labeled Indole-3-carboxaldehyde. It is a biochemical used to prepare analogs of the indole phytoalexin cyclobrassinin. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, primarily as tracers for quantitation during the drug development process. Deuteration has gained attention due to its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • For research use only
  • Incorporates stable heavy isotopes for quantitation in drug development