Learn More
Medchemexpress LLC Indole-3-carboxylic acid | 771-50-6 | 100.0% | 161.16 | 1 ML
SDP
Catalog No. 5000404673
Encompass_Preferred
Indole-3-carboxylic acid is an orally active urinary indolic tryptophan metabolite. It mediates priming against *Plectosphaerella cucumerina* and enhances the anti-colorectal cancer potency of Doxorubicin by inducing cell senescence. It can also be utilized in liver disease research.
  • Orally active urinary indolic tryptophan metabolite.
  • Mediator of priming against *Plectosphaerella cucumerina*.
  • Enhances anti-colorectal cancer potency of Doxorubicin through cell senescence induction.
  • Applicable in liver disease research.
  • For research use only.
Catalog No. 50-004-04673 Supplier Medchemexpress LLC Supplier No. HY4016110MM/1ML
Indole-3-carboxylic acid is an orally active urinary indolic tryptophan metabolite. It mediates priming against *Plectosphaerella cucumerina* and enhances the anti-colorectal cancer potency of Doxorubicin by inducing cell senescence. It can also be utilized in liver disease research.
  • Orally active urinary indolic tryptophan metabolite.
  • Mediator of priming against *Plectosphaerella cucumerina*.
  • Enhances anti-colorectal cancer potency of Doxorubicin through cell senescence induction.
  • Applicable in liver disease research.
  • For research use only.