Learn More
Medchemexpress LLC Indole-3-carboxylic acid | 771-50-6 | 161.16 | 10 G
SDP
Catalog No. 5000405135
Encompass_Preferred
Indole-3-carboxylic acid is an orally active urinary indolic tryptophan metabolite. It acts as a mediator of priming against *Plectosphaerella cucumerina* and enhances the anti-colorectal cancer potency of Doxorubicin by inducing cell senescence. It can also be used in liver disease research.
  • Orally active urinary indolic tryptophan metabolite.
  • Mediates priming against *Plectosphaerella cucumerina*.
  • Enhances anti-colorectal cancer potency by inducing cell senescence.
  • Can be used in liver disease research.
Catalog No. 50-004-05135 Supplier Medchemexpress LLC Supplier No. HY4016110G
Indole-3-carboxylic acid is an orally active urinary indolic tryptophan metabolite. It acts as a mediator of priming against *Plectosphaerella cucumerina* and enhances the anti-colorectal cancer potency of Doxorubicin by inducing cell senescence. It can also be used in liver disease research.
  • Orally active urinary indolic tryptophan metabolite.
  • Mediates priming against *Plectosphaerella cucumerina*.
  • Enhances anti-colorectal cancer potency by inducing cell senescence.
  • Can be used in liver disease research.