Learn More
Medchemexpress LLC Indole-3-carboxylic acid | 771-50-6 | 99.6% | 161.16 | 5 MG
SDP
Catalog No. 5000405146
Encompass_Preferred
Indole-3-carboxylic acid (Standard) is the analytical standard of Indole-3-carboxylic acid. It is intended for research and analytical applications. Indole-3-carboxylic acid is a normal urinary indolic tryptophan metabolite and has been found to be elevated in patients with liver diseases. The compound is of analytical standard grade, used as a reference in qualitative, quantitative, and methodological research experiments, particularly in HPLC, GC, and MS.
  • Analytical standard for Indole-3-carboxylic acid.
  • Intended for research and analytical applications.
  • Used in qualitative, quantitative, and methodological research experiments (HPLC, GC, MS).
Catalog No. 50-004-05146 Supplier Medchemexpress LLC Supplier No. HY40161R5MG
Indole-3-carboxylic acid (Standard) is the analytical standard of Indole-3-carboxylic acid. It is intended for research and analytical applications. Indole-3-carboxylic acid is a normal urinary indolic tryptophan metabolite and has been found to be elevated in patients with liver diseases. The compound is of analytical standard grade, used as a reference in qualitative, quantitative, and methodological research experiments, particularly in HPLC, GC, and MS.
  • Analytical standard for Indole-3-carboxylic acid.
  • Intended for research and analytical applications.
  • Used in qualitative, quantitative, and methodological research experiments (HPLC, GC, MS).