Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Indole-3-carboxylic acid | 771-50-6 | 99.96% | 161.16 | 1 G
SDP

Catalog No. 5000405136
Encompass_Preferred

Indole-3-carboxylic acid is an orally active urinary indolic tryptophan metabolite. This compound mediates priming against Plectosphaerella cucumerina and enhances anti-colorectal cancer potency by inducing cell senescence. It is also applicable in liver disease research.

  • High purity of 99.96%.
  • Orally active urinary indolic tryptophan metabolite.
  • Mediates priming against Plectosphaerella cucumerina.
  • Enhances anti-colorectal cancer effects via cell senescence.
  • Applicable in liver disease research.
  • Available as a 10 mM * 1 mL solution in DMSO.
  • Available in solid forms: 1 g, 5 g, 10 g, 25 g, 50 g, and > 100 g.

Catalog No. 50-004-05136 Supplier Medchemexpress LLC Supplier No. HY401611G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Indole-3-carboxylic acid is an orally active urinary indolic tryptophan metabolite. This compound mediates priming against Plectosphaerella cucumerina and enhances anti-colorectal cancer potency by inducing cell senescence. It is also applicable in liver disease research.

  • High purity of 99.96%.
  • Orally active urinary indolic tryptophan metabolite.
  • Mediates priming against Plectosphaerella cucumerina.
  • Enhances anti-colorectal cancer effects via cell senescence.
  • Applicable in liver disease research.
  • Available as a 10 mM * 1 mL solution in DMSO.
  • Available in solid forms: 1 g, 5 g, 10 g, 25 g, 50 g, and > 100 g.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.