Learn More
Medchemexpress LLC Indole-3-carboxylic acid | 771-50-6 | 99.96% | 161.16 | 25 G
SDP
Catalog No. 5000402182
Encompass_Preferred
Indole-3-carboxylic acid is an orally active urinary indolic tryptophan metabolite. It mediates priming against *Plectosphaerella cucumerina* and enhances the anti-colorectal cancer potency of Doxorubicin by inducing cell senescence. It can also be used in liver disease research.
  • Mediates priming against *Plectosphaerella cucumerina*.
  • Enhances the anti-colorectal cancer potency of Doxorubicin by inducing cell senescence.
  • Can be used in liver disease research.
  • For research use only.
  • Powder storage: -20°C for 3 years, 4°C for 2 years.
  • In solvent storage: -80°C for 6 months, -20°C for 1 month.
Catalog No. 50-004-02182 Supplier Medchemexpress LLC Supplier No. 25GHY40161
Indole-3-carboxylic acid is an orally active urinary indolic tryptophan metabolite. It mediates priming against *Plectosphaerella cucumerina* and enhances the anti-colorectal cancer potency of Doxorubicin by inducing cell senescence. It can also be used in liver disease research.
  • Mediates priming against *Plectosphaerella cucumerina*.
  • Enhances the anti-colorectal cancer potency of Doxorubicin by inducing cell senescence.
  • Can be used in liver disease research.
  • For research use only.
  • Powder storage: -20°C for 3 years, 4°C for 2 years.
  • In solvent storage: -80°C for 6 months, -20°C for 1 month.