Learn More
Medchemexpress LLC Indole-3-carboxylic acid | 771-50-6 | 99.96% | 161.16 | 5 G
SDP
Catalog No. 5000401596
Encompass_Preferred
Indole-3-carboxylic acid is an orally active urinary indolic tryptophan metabolite. It mediates priming against *Plectosphaerella cucumerina* and enhances the anti-colorectal cancer potency of Doxorubicin by inducing cell senescence. It can also be utilized in liver disease research.
  • Orally active urinary indolic tryptophan metabolite.
  • Mediates priming against *Plectosphaerella cucumerina*.
  • Enhances anti-colorectal cancer potency by inducing cell senescence.
  • Can be used in liver disease research.
  • High purity of 99.96%.
  • Available in solid form.
  • Soluble in DMSO.
  • Multiple in vivo dissolution methods available.
Catalog No. 50-004-01596 Supplier Medchemexpress LLC Supplier No. HY401615G
Indole-3-carboxylic acid is an orally active urinary indolic tryptophan metabolite. It mediates priming against *Plectosphaerella cucumerina* and enhances the anti-colorectal cancer potency of Doxorubicin by inducing cell senescence. It can also be utilized in liver disease research.
  • Orally active urinary indolic tryptophan metabolite.
  • Mediates priming against *Plectosphaerella cucumerina*.
  • Enhances anti-colorectal cancer potency by inducing cell senescence.
  • Can be used in liver disease research.
  • High purity of 99.96%.
  • Available in solid form.
  • Soluble in DMSO.
  • Multiple in vivo dissolution methods available.