Learn More
Bioworld Indole-3-Carboxylic Acid (ICA), 10 g
Catalog No. 50255030
Encompass_Preferred
Specification:Purity: >99%Melting point (°C): 224-230
Catalog No. 50-255-030 Supplier Bioworld Supplier No. 306310002
Specification:Purity: >99%Melting point (°C): 224-230