Learn More
Medchemexpress LLC Indole-3-carboxylic acid (standard) | 771-50-6 | 99.58% | 161.16 | 100 MG
SDP
Catalog No. 5000403916
Encompass_Preferred
Indole-3-carboxylic acid (Standard) is the analytical standard of Indole-3-carboxylic acid and is intended for research and analytical applications. It is a normal urinary indolic tryptophan metabolite that has been found to be elevated in patients with liver diseases. This product is for research use only and is not sold to patients.
  • Analytical standard for research and analytical applications
  • Normal urinary indolic tryptophan metabolite
  • Found elevated in patients with liver diseases
  • Commonly used in qualitative, quantitative, and methodological research experiments
  • Suitable for HPLC, GC, and MS
Catalog No. 50-004-03916 Supplier Medchemexpress LLC Supplier No. HY40161R100MG
Indole-3-carboxylic acid (Standard) is the analytical standard of Indole-3-carboxylic acid and is intended for research and analytical applications. It is a normal urinary indolic tryptophan metabolite that has been found to be elevated in patients with liver diseases. This product is for research use only and is not sold to patients.
  • Analytical standard for research and analytical applications
  • Normal urinary indolic tryptophan metabolite
  • Found elevated in patients with liver diseases
  • Commonly used in qualitative, quantitative, and methodological research experiments
  • Suitable for HPLC, GC, and MS