Learn More
Medchemexpress LLC Indole-3-carboxylic acid (Standard) | 771-50-6 | 99.58% | 161.16 | 25 MG
SDP
Catalog No. 5000402472
Encompass_Preferred
Indole-3-carboxylic acid (Standard) is the analytical standard of Indole-3-carboxylic acid. It is intended for research and analytical applications. Indole-3-carboxylic acid is a normal urinary indolic tryptophan metabolite and has been found elevated in patients with liver diseases.
  • The compound is an analytical standard, commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Appearance: solid, off-white to light yellow.
  • Initial source: endogenous metabolite, human gut microbiota metabolites.
  • Shipping: room temperature in continental US; may vary elsewhere.
Catalog No. 50-004-02472 Supplier Medchemexpress LLC Supplier No. 25MGHY40161R
Indole-3-carboxylic acid (Standard) is the analytical standard of Indole-3-carboxylic acid. It is intended for research and analytical applications. Indole-3-carboxylic acid is a normal urinary indolic tryptophan metabolite and has been found elevated in patients with liver diseases.
  • The compound is an analytical standard, commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Appearance: solid, off-white to light yellow.
  • Initial source: endogenous metabolite, human gut microbiota metabolites.
  • Shipping: room temperature in continental US; may vary elsewhere.