Learn More
Medchemexpress LLC Indole-3-carboxylic acid (standard) | 771-50-6 | 99.6% | 161.16 | 50 MG
SDP
Catalog No. 5000405131
Encompass_Preferred
Indole-3-carboxylic acid (Standard) is the analytical standard of Indole-3-carboxylic acid. This product is intended for research and analytical applications. It is a normal urinary indolic tryptophan metabolite found elevated in patients with liver diseases.
  • Used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS.
  • Analytical standard for research and analytical applications.
  • Reference standard supplied assay.
Catalog No. 50-004-05131 Supplier Medchemexpress LLC Supplier No. HY40161R50MG
Indole-3-carboxylic acid (Standard) is the analytical standard of Indole-3-carboxylic acid. This product is intended for research and analytical applications. It is a normal urinary indolic tryptophan metabolite found elevated in patients with liver diseases.
  • Used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS.
  • Analytical standard for research and analytical applications.
  • Reference standard supplied assay.