Learn More
Medchemexpress LLC Indole-3-lactate-O-β-D-glucopyranoside | 143884-93-9 | 99.9% | 395.40 g·mol⁻¹ | C19H25NO8 | 10 MG
SDP
Catalog No. 5000295751
Encompass_Preferred
Indole-3-lactate-O-β-D-glucopyranoside is an endogenous plant metabolite used as a research reagent and plant growth regulator. It is supplied as a white to off-white solid with high purity and good aqueous solubility, intended for biochemical and plant biology studies.
  • High purity suitable for research applications.
  • White to off-white solid form for easy handling and storage.
  • Soluble in water (≈125 mg/mL) with sonication if needed.
  • Stable as powder at -20°C for long-term storage.
  • Available in small research pack sizes for lab-scale studies.
Catalog No. 50-002-95751 Supplier Medchemexpress LLC Supplier No. HYN1329510MG
Indole-3-lactate-O-β-D-glucopyranoside is an endogenous plant metabolite used as a research reagent and plant growth regulator. It is supplied as a white to off-white solid with high purity and good aqueous solubility, intended for biochemical and plant biology studies.
  • High purity suitable for research applications.
  • White to off-white solid form for easy handling and storage.
  • Soluble in water (≈125 mg/mL) with sonication if needed.
  • Stable as powder at -20°C for long-term storage.
  • Available in small research pack sizes for lab-scale studies.