Learn More
Cayman Chemical Indole-3-lactIc AcId 100mg
SDP
Catalog No. 502916153
Encompass_Preferred
A metabolite of tryptophan; inhibits LPS-induced production of IL-8 in Caco-2 cells at 0.1-10 mM; reduces colon tumor number and size in cancer-prone Apc+/Min mice at 20 mg/kg
Catalog No. 50-291-6153 Supplier Cayman Chemical Supplier No. 37534100 MG
A metabolite of tryptophan; inhibits LPS-induced production of IL-8 in Caco-2 cells at 0.1-10 mM; reduces colon tumor number and size in cancer-prone Apc+/Min mice at 20 mg/kg