Learn More
Medchemexpress LLC Indole-3-lactic acid | 1821-52-9 | MFCD00005642 | 100.0% | 205.21 g·mol⁻1 | C11H11NO3 | 5 MG
SDP
Catalog No. 5000262997
Encompass_Preferred
Indole-3-lactic acid is an endogenous tryptophan metabolite supplied as an analytical reference standard for research and analytical applications. The compound is an off-white to light-yellow solid (CAS 1821-52-9) with a molecular formula of C11H11NO3 and molecular weight 205.21 g·mol⁻1; it is intended for method development, calibration, and metabolite identification.
Catalog No. 50-002-62997 Supplier Medchemexpress LLC Supplier No. HY113099R5MG
Indole-3-lactic acid is an endogenous tryptophan metabolite supplied as an analytical reference standard for research and analytical applications. The compound is an off-white to light-yellow solid (CAS 1821-52-9) with a molecular formula of C11H11NO3 and molecular weight 205.21 g·mol⁻1; it is intended for method development, calibration, and metabolite identification.