Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Indole-3-methanamine | 22259-53-6 | 146.19 | 1 G
SDP

Catalog No. 5000401486
Encompass_Preferred

Indole-3-methanamine is identified as a potential biomarker for the consumption of foods such as barley, cereals, and cereal products. This product is supplied as a solid for research and analytical applications.

  • Store at -20°C, protected from light.
  • When in solvent, store at -80°C for up to 6 months or -20°C for up to 1 month, protected from light.
  • Soluble in DMSO at 50 mg/mL.
  • Melting point: 101-102°C.
  • Boiling point: 335.6°C at 760 mmHg.
  • Flash point: 183°C.
  • Relative density: 1.199 g/cm³.

Catalog No. 50-004-01486 Supplier Medchemexpress LLC Supplier No. HY338931G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Indole-3-methanamine is identified as a potential biomarker for the consumption of foods such as barley, cereals, and cereal products. This product is supplied as a solid for research and analytical applications.

  • Store at -20°C, protected from light.
  • When in solvent, store at -80°C for up to 6 months or -20°C for up to 1 month, protected from light.
  • Soluble in DMSO at 50 mg/mL.
  • Melting point: 101-102°C.
  • Boiling point: 335.6°C at 760 mmHg.
  • Flash point: 183°C.
  • Relative density: 1.199 g/cm³.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.