Learn More
Medchemexpress LLC Indole-3-pyruvic acid | 392-12-1 | MFCD00005640 | 99.6% | 203.19 g/mol | C11H9NO3 | 1 ML
SDP
Catalog No. 5000300171
Encompass_Preferred
Indole-3-pyruvic acid is an endogenous tryptophan metabolite and aryl hydrocarbon receptor (AHR) agonist supplied as a solid or a ready-to-use 10 mM solution in DMSO for biochemical and in vitro research. The material is provided with validated purity, molecular weight, and solubility data to support experimental reproducibility.
  • High purity suitable for research applications.
  • Ready-to-use 10 mM solution in DMSO for rapid assay setup.
  • Available as solid for custom dissolution and storage.
  • Documented solubility in DMSO and water with handling recommendations.
  • Molecular weight and formula supplied for dosing and calculations.
  • Appropriate for biochemical and pharmacological assays.
Catalog No. 50-003-00171 Supplier Medchemexpress LLC Supplier No. HYW02839310MM/1ML
Indole-3-pyruvic acid is an endogenous tryptophan metabolite and aryl hydrocarbon receptor (AHR) agonist supplied as a solid or a ready-to-use 10 mM solution in DMSO for biochemical and in vitro research. The material is provided with validated purity, molecular weight, and solubility data to support experimental reproducibility.
  • High purity suitable for research applications.
  • Ready-to-use 10 mM solution in DMSO for rapid assay setup.
  • Available as solid for custom dissolution and storage.
  • Documented solubility in DMSO and water with handling recommendations.
  • Molecular weight and formula supplied for dosing and calculations.
  • Appropriate for biochemical and pharmacological assays.