Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Indole-3-pyruvic acid | 392-12-1 | MFCD00005640 | 99.6% | 203.19 g·mol⁻¹ | C11H9NO3 | 100 MG
SDP

Catalog No. 5000299863
Encompass_Preferred

Indole-3-pyruvic acid is an endogenous tryptophan metabolite and keto analogue that functions as an aryl hydrocarbon receptor (AHR) agonist with reported antioxidant activity. It is supplied as a research-grade chemical for biochemical and pharmacological studies.

  • Endogenous metabolite and keto analogue of tryptophan.
  • Acts as an aryl hydrocarbon receptor (AHR) agonist.
  • Exhibits antioxidant activity in reported studies.
  • High purity (99.55%).
  • Molecular formula C11H9NO3; molecular weight 203.19 g·mol⁻¹.
  • Available in small research sizes, including 100 mg, and DMSO solution formats.
  • Soluble in DMSO and water.

Catalog No. 50-002-99863 Supplier Medchemexpress LLC Supplier No. HYW028393100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Indole-3-pyruvic acid is an endogenous tryptophan metabolite and keto analogue that functions as an aryl hydrocarbon receptor (AHR) agonist with reported antioxidant activity. It is supplied as a research-grade chemical for biochemical and pharmacological studies.

  • Endogenous metabolite and keto analogue of tryptophan.
  • Acts as an aryl hydrocarbon receptor (AHR) agonist.
  • Exhibits antioxidant activity in reported studies.
  • High purity (99.55%).
  • Molecular formula C11H9NO3; molecular weight 203.19 g·mol⁻¹.
  • Available in small research sizes, including 100 mg, and DMSO solution formats.
  • Soluble in DMSO and water.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.