Learn More
Medchemexpress LLC Indole-3-pyruvic acid | 392-12-1 | MFCD00005640 | 99.6% | 203.19 g/mol | C11H9NO3 | 50 MG
SDP
Catalog No. 5000300766
Encompass_Preferred
Indole-3-pyruvic acid is an endogenous metabolite and keto analogue of tryptophan used as a research reagent and analytical standard in studies of AHR activation, inflammation, and oxidative stress. It is supplied as a high-purity chemical for laboratory use.
  • High purity suitable for analytical and biochemical applications.
  • Endogenous metabolite and AHR agonist for pathway studies.
  • Supplied in research-scale quantities for laboratory testing.
  • Verified molecular formula and molecular weight for reference.
  • Useful as a reagent in inflammation and oxidative stress research.
Catalog No. 50-003-00766 Supplier Medchemexpress LLC Supplier No. HYW02839350MG
Indole-3-pyruvic acid is an endogenous metabolite and keto analogue of tryptophan used as a research reagent and analytical standard in studies of AHR activation, inflammation, and oxidative stress. It is supplied as a high-purity chemical for laboratory use.
  • High purity suitable for analytical and biochemical applications.
  • Endogenous metabolite and AHR agonist for pathway studies.
  • Supplied in research-scale quantities for laboratory testing.
  • Verified molecular formula and molecular weight for reference.
  • Useful as a reagent in inflammation and oxidative stress research.