Learn More
Medchemexpress LLC Indole-5-carboxylic acid | 1670-81-1 | 99.6% | C9H7NO2 | 5 G
SDP
Catalog No. 5000433985
Encompass_Preferred
Indole-5-carboxylic acid is an indole derivative. It is intended for research use only and not sold to patients.
  • Inhibits corrosion of low carbon steel
  • Effective in deoxygenated 0.5 M sulfuric acid solution
Catalog No. 50-004-33985 Supplier Medchemexpress LLC Supplier No. HY593025G
Indole-5-carboxylic acid is an indole derivative. It is intended for research use only and not sold to patients.
  • Inhibits corrosion of low carbon steel
  • Effective in deoxygenated 0.5 M sulfuric acid solution