Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Indole-6-carboxylic Acid 98.0+%, TCI America™
SDP

Catalog No. I06635G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I06635G 5 g
I06631G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I06635G Supplier TCI America Supplier No. I06635G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1670-82-2
Molecular Formula C9H7NO2
Molecular Weight (g/mol) 161.16
MDL Number MFCD00210441
InChI Key GHTDODSYDCPOCW-UHFFFAOYSA-N
Synonym indole-6-carboxylic acid, 6-indolecarboxylic acid, 6-carboxyindole, 6-indole carboxylic acid, pubchem1695, 6-carboxy-1h-indole, indole-6-carboxylicacid, indol-6-carboxylic acid, indole 6-carboxylic acid, maybridge4_003509
PubChem CID 595230
IUPAC Name 1H-indole-6-carboxylic acid
SMILES C1=CC(=CC2=C1C=CN2)C(=O)O

Specifications

Melting Point 257°C
Quantity 5 g
Formula Weight 161.16
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Indole-6-carboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.