Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich indole-7-methanol

Catalog No. CDS008595 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS008595 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS008595 Supplier Sigma Aldrich Supplier No. CDS008595100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Biotransformation of 2,3-dihydrobenzofuran using intact cells of pseudomonas putida uv4 has been investigated
  • 2,3-dihydrobenzofuran is the intermediate formed during catalytic hydrodeoxygenation of benzofuran

Specifications

CAS 1074-87-9
Molecular Formula C9H9NO
Linear Formula C9H9NO
Quantity 5 g
MDL Number MFCD02179597
Molecular Weight (g/mol) 147.17
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.