Learn More
Sigma Aldrich Fine Chemicals Biosciences Indole Assay Kit sufficien
Catalog No. 502900015
Encompass
Indole is the primary product of tryptophan breakdown by tryptophanase. The indole test is commonly performed on bacteria to classify them on their ability to break down tryptophan to indole.
Catalog No. 50-290-0015 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. MAK3261KT
Add to Cart
Add to Cart
Indole is the primary product of tryptophan breakdown by tryptophanase. The indole test is commonly performed on bacteria to classify them on their ability to break down tryptophan to indole.