Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Indole-C2-amide-C2-NH2 | 215789-29-0 | 99.4% | 231.29 | 1 ML
SDP

Catalog No. 5000359688
Encompass_Preferred

Indole-C2-amide-C2-NH2 is a potent linker. This viscous, light yellow to yellow liquid is stable under recommended storage conditions, making it suitable for various chemical synthesis applications.

  • Potent linker
  • Viscous liquid, light yellow to yellow appearance
  • Stable under recommended storage conditions
  • Purity of 99.42% by LCMS
  • Pure form storage: up to 3 years at -20°C or 2 years at 4°C
  • In solvent storage: up to 6 months at -80°C or 1 month at -20°C

Catalog No. 50-003-59688 Supplier Medchemexpress LLC Supplier No. HY14808010MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Indole-C2-amide-C2-NH2 is a potent linker. This viscous, light yellow to yellow liquid is stable under recommended storage conditions, making it suitable for various chemical synthesis applications.

  • Potent linker
  • Viscous liquid, light yellow to yellow appearance
  • Stable under recommended storage conditions
  • Purity of 99.42% by LCMS
  • Pure form storage: up to 3 years at -20°C or 2 years at 4°C
  • In solvent storage: up to 6 months at -80°C or 1 month at -20°C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.