Learn More
Medchemexpress LLC Indole-d6 | 104959-27-5 | 123.18 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYW001132S10MG
Encompass_Preferred
Indole-d6 is a deuterium-labeled indole and an endogenous metabolite. It is intended for research use only.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Store powder at -20°C for 3 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month
Catalog No. 50-004-47109