Learn More
Medchemexpress LLC Indole-d6 | 104959-27-5 | MFCD22124219 | 123.18 | 5 MG
SDP
Catalog No. 5000449174
Encompass_Preferred
Indole-d6 is the deuterium labeled Indole, an endogenous metabolite. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-49174 Supplier Medchemexpress LLC Supplier No. HYW001132S5MG
Indole-d6 is the deuterium labeled Indole, an endogenous metabolite. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.