Learn More
Medchemexpress LLC Indole-d7 | 73509-20-3 | 124.19 | 10 MG
SDP
Catalog No. 5000447099
Encompass_Preferred
Indole-d7 is a deuterium-labeled form of Indole, an endogenous metabolite. This product is intended for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration may influence the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-47099 Supplier Medchemexpress LLC Supplier No. HYW001132S410MG
Indole-d7 is a deuterium-labeled form of Indole, an endogenous metabolite. This product is intended for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration may influence the pharmacokinetic and metabolic profiles of drugs.