Learn More
Medchemexpress LLC Indole-d7 | 73509-20-3 | 124.19 | 100 MG
SDP
Catalog No. 5000446316
Encompass_Preferred
Indole-d7 is a deuterium-labeled indole that serves as an endogenous metabolite. Deuteration can impact the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-46316 Supplier Medchemexpress LLC Supplier No. HYW001132S4100MG
Indole-d7 is a deuterium-labeled indole that serves as an endogenous metabolite. Deuteration can impact the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS