Learn More
Medchemexpress LLC Indole-d7 | 73509-20-3 | 124.19 | 25 MG
SDP
Catalog No. 5000445402
Encompass_Preferred
Indole-d7 is a deuterium labeled indole, which is an endogenous metabolite. It is for research use only. This compound can be used as a tracer and as an internal standard for quantitative analysis.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-45402 Supplier Medchemexpress LLC Supplier No. 25MGHYW001132S4
Indole-d7 is a deuterium labeled indole, which is an endogenous metabolite. It is for research use only. This compound can be used as a tracer and as an internal standard for quantitative analysis.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuteration has the potential to affect the pharmacokinetic and metabolic profiles of drugs