Learn More
Medchemexpress LLC Indole-d7 | 73509-20-3 | 124.19 | 50 MG
SDP
Catalog No. 5000449742
Encompass_Preferred
Indole-d7 is a deuterium-labeled form of Indole, which is an endogenous metabolite. It serves as a tracer and an internal standard for quantitative analysis methods such as NMR, GC-MS, or LC-MS. The incorporation of stable heavy isotopes, like deuterium, into drug molecules is primarily used for quantitation during drug development processes, with potential effects on pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuterium labeling can affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-49742 Supplier Medchemexpress LLC Supplier No. HYW001132S450MG
Add to Cart
Add to Cart
Indole-d7 is a deuterium-labeled form of Indole, which is an endogenous metabolite. It serves as a tracer and an internal standard for quantitative analysis methods such as NMR, GC-MS, or LC-MS. The incorporation of stable heavy isotopes, like deuterium, into drug molecules is primarily used for quantitation during drug development processes, with potential effects on pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuterium labeling can affect pharmacokinetic and metabolic profiles of drugs