Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Indole, MilliporeSigma™ Supelco™

Catalog No. 111020580 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1000 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0580 1000 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-0580 Supplier MilliporeSigma Supelco Supplier No. 442619

Indole is classified under the volatile flavor compounds (VFCs). It is known to play significant role in various biological functions such as anti-inflammatory, anticonvulsant, cardiovascular and antibacterial activities.

Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampule
CAS 120-72-9
Flash Point 121°C
Physical Form Neat
Linear Formula C8H7 N
MDL Number MFCD00005607
Melting Point 51°C to 54°C
Boiling Point 253°C to 254°C
Synonym 1 H-Benzo[b]pyrrole
Type Food and Cosmetic Component Standard
Quantity 1000 mg
Formula Weight 117.15
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.