Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Indole (Off-white Crystalline Solid/Certified), Fisher Chemical

Catalog No. O610425 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
O6104-25 25 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O6104-25

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 120-72-9
Molecular Formula C8H7N
Molecular Weight (g/mol) 117.151
MDL Number MFCD00005607
InChI Key SIKJAQJRHWYJAI-UHFFFAOYSA-N
Synonym indole, indol, 2,3-benzopyrrole, 1-benzazole, ketole, 1-azaindene, benzopyrrole, 2,3-benzopyrole, caswell no. 498b, indol german
PubChem CID 798
ChEBI CHEBI:16881
IUPAC Name 1H-indole
SMILES C1=CC=C2C(=C1)C=CN2

Specifications

Melting Point 51°C
Color White
Density 1.220g/cm³
pH 5.9
Boiling Point 253°C
Infrared Spectrum Conforms to reference
Assay Percent Range ≥98.0 %
Packaging Glass Bottle
Quantity 25 g
Vapor Pressure 0.012mmHg at 25°C
Formula Weight 117.15
Percent Purity ≥98.0%
Grade Certified
Physical Form Solid
Chemical Name or Material Indole
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.