Learn More
Medchemexpress LLC Indole (standard) | 120-72-9 | 117.15 | 10 MG
SDP
Catalog No. 5000447570
Encompass_Preferred
Indole (Standard) is the analytical standard of Indole, intended for research and analytical applications. It is an aromatic, heterocyclic, organic compound widely distributed in nature and produced by various bacteria. Indole regulates bacterial physiology, including spore formation, plasmid stability, drug resistance, biofilm formation, and virulence, acting as an intercellular signal molecule.
  • Grade of analytical standard
  • Reference standard supplied for assay
  • Commonly used in qualitative, quantitative, and methodological research experiments
  • Suitable for HPLC, GC, and MS applications
Catalog No. 50-004-47570 Supplier Medchemexpress LLC Supplier No. HYW001132R10MG
Indole (Standard) is the analytical standard of Indole, intended for research and analytical applications. It is an aromatic, heterocyclic, organic compound widely distributed in nature and produced by various bacteria. Indole regulates bacterial physiology, including spore formation, plasmid stability, drug resistance, biofilm formation, and virulence, acting as an intercellular signal molecule.
  • Grade of analytical standard
  • Reference standard supplied for assay
  • Commonly used in qualitative, quantitative, and methodological research experiments
  • Suitable for HPLC, GC, and MS applications