Learn More
Medchemexpress LLC Indole (standard) | 120-72-9 | 117.15 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW001132R100MG
Encompass_Preferred
Indole (Standard) is the analytical standard of Indole, intended for research and analytical applications. It is an aromatic, heterocyclic, organic compound widely distributed in the natural environment and produced by various bacteria. It regulates bacterial physiology, including spore formation, plasmid stability, drug resistance, biofilm formation, and virulence as an intercellular signal molecule.
  • Used as a reference standard for assays.
  • Commonly used in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
  • Regulates bacterial physiology, including spore formation, plasmid stability, resistance to drugs, biofilm formation, and virulence.
  • Widely distributed in the natural environment.
Catalog No. 50-004-46914