Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Indole (standard) | 120-72-9 | 117.15 | 250 MG
SDP

Catalog No. 5000450037
Encompass_Preferred

Indole (standard) is the analytical standard of Indole, intended for research and analytical applications. It is an aromatic, heterocyclic, organic compound widely distributed in the natural environment and produced by various bacteria. Indole regulates bacterial physiology, including spore formation, plasmid stability, resistance to drugs, biofilm formation, and virulence as an intercellular signal molecule.

  • Application: The compound is an analytical standard used as a reference standard in assays. It is commonly utilized in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Initial source: Endogenous metabolite, human gut microbiota metabolites, microorganisms.
  • Appearance: Solid
  • Color: Off-white to light yellow.

Catalog No. 50-004-50037 Supplier Medchemexpress LLC Supplier No. 250MGHYW001132R
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Indole (standard) is the analytical standard of Indole, intended for research and analytical applications. It is an aromatic, heterocyclic, organic compound widely distributed in the natural environment and produced by various bacteria. Indole regulates bacterial physiology, including spore formation, plasmid stability, resistance to drugs, biofilm formation, and virulence as an intercellular signal molecule.

  • Application: The compound is an analytical standard used as a reference standard in assays. It is commonly utilized in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Initial source: Endogenous metabolite, human gut microbiota metabolites, microorganisms.
  • Appearance: Solid
  • Color: Off-white to light yellow.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.