Learn More
Medchemexpress LLC Indole (standard) | 120-72-9 | 117.15 | 50 MG
SDP
Catalog No. 5000447111
Encompass_Preferred
Indole (Standard) is an analytical standard, an aromatic, heterocyclic, organic compound found widely in nature and produced by various bacteria. It acts as an intercellular signal molecule, regulating bacterial physiology such as spore formation, plasmid stability, drug resistance, biofilm formation, and virulence. It is intended for research and analytical applications, commonly utilized in qualitative, quantitative, and methodological research experiments.
  • Used as an analytical standard
  • Regulates bacterial physiology
  • Acts as an intercellular signal molecule
  • Applied in HPLC, GC, and MS experiments
Catalog No. 50-004-47111 Supplier Medchemexpress LLC Supplier No. HYW001132R50MG
Indole (Standard) is an analytical standard, an aromatic, heterocyclic, organic compound found widely in nature and produced by various bacteria. It acts as an intercellular signal molecule, regulating bacterial physiology such as spore formation, plasmid stability, drug resistance, biofilm formation, and virulence. It is intended for research and analytical applications, commonly utilized in qualitative, quantitative, and methodological research experiments.
  • Used as an analytical standard
  • Regulates bacterial physiology
  • Acts as an intercellular signal molecule
  • Applied in HPLC, GC, and MS experiments