Learn More
Medchemexpress LLC Indolelactic acid-d5 | 2470130-19-7 | 98.2% | 210.24 | 1 MG
SDP
Catalog No. 5000438434
Encompass_Preferred
Indolelactic acid-d5 is the deuterium labeled Indolelactic acid. Indolelactic acid is a tryptophan (Trp) catabolite in *Azotobacter vinelandii* cultures. It is for research use only and not sold to patients.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-38434 Supplier Medchemexpress LLC Supplier No. HY113099S1MG
Indolelactic acid-d5 is the deuterium labeled Indolelactic acid. Indolelactic acid is a tryptophan (Trp) catabolite in *Azotobacter vinelandii* cultures. It is for research use only and not sold to patients.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.